Learn More
VCP (Valosin Containing Protein) Mouse, Unlabeled, Clone: 18, BD
Mouse Monoclonal Antibody
Marque: BD Biosciences 612183
Informations supplémentaires : Poids : 0.01000kg
Description
ATPases that belong to the ATPase associated with different cellular activities (AAA) family are homo-oligomeric proteins that have roles in cell cycle regulation, protein degradation, organelle biogenesis, and vesicle mediated-protein transport. Valosin containing protein (VCP) is a AAA family member that is found as a hexamer in rat liver. In vitro, VCP catalyzes the hydrolysis of ATP, but this activity is distinct from classical transport ATPases. VCP protein is found primarily in the transitional elements between the rough and smooth ER (TER), but stimulation with EGF leads to VCP translocation to the nucleus. Antibodies to VCP can perturb cell-free formation of transition vesicles from isolated TER of rat liver, and can inhibit transfer of material from ER to Golgi in a reconstituted membrane system. VCP is phosphorylated after T-cell activation, and PTPH1 dephosphorylation of VCP correlates with suppression of cell growth. In addition, the N-terminal region of VCP binds to the DNA damage repair protein, BRCA1. Thus, VCP may have roles in both ER protein transport and nuclear function, which are important for cell growth and survival.
Immunofluorescence, Western Blotting
Spécification
VCP (Valosin Containing Protein) | |
Monoclonal | |
250μg/mL | |
Valosin Containing Protein | |
Mouse | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at -20°C. |
Western Blot | |
18 | |
Unconjugated | |
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
Mouse VCP aa. 9-130 | |
150 μg | |
Cell Biology | |
Canine, Chicken, Human, Mouse, Rat | |
IgG1 |
For Research Use Only.