missing translation for 'onlineSavingsMsg'
Learn More
Learn More
APC4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 14129-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
Description
A large protein complex, termed the anaphase-promoting complex (APC), or the cyclosome, promotes metaphase-anaphase transition by ubiquitinating its specific substrates such as mitotic cyclins and anaphase inhibitor, which are subsequently degraded by the 26S proteasome. Biochemical studies have shown that the vertebrate APC contains eight subunits. The composition of the APC is highly conserved in organisms from yeast to humans. The exact function of this gene product is not known. Two transcript variants encoding different isoforms have been found for this gene.Spécification
APC4 | |
0.15 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q91W96, Q9UJX5 | |
ANAPC4, APC4, Cyclosome subunit 4 | |
Rabbit | |
IgG | |
150 μL | |
-20°C | |
Primary | |
29945, 305420, 52206 |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
ANAPC4 | |
Liquid | |
Anapc4 | |
APC4 Fusion Protein Ag5225 | |
Antigen Affinity Chromatography | |
RUO | |
Antibody | |
Polyclonal | |
Human, Mouse, Rat |