missing translation for 'onlineSavingsMsg'
Learn More

Apolipoprotein A II Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marque:  Proteintech 16845-1-AP-150UL

Informations supplémentaires : Poids : 0.00010kg

 Afficher plus de versions de ce produit

Code produit. 16806614

  • 329.85€ / 150µL

Expédition estimée
10-03-2021

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

At least 9 distinct polymorphic forms of apolipoproteins are known. The apolipoproteins act as stabilizers of the intact lipoprotein particles. Quantitative measurements of HDL, LDL, and VLDL particles of human serum are often used to estimate an individual′s relative risk of coronary heart disease. In addition, quantitative immunological measurements of certain apolipoproteins ( especially A-1 and B) have been suggested to be more accurate estimators of coronary heart disease that measurements of lipoprotein particles (especially HDL and LDL). The apolipoproteins are derived from human plasma by density gradient ultracentrifugation and HPLC.
Spécification

Spécification

Apolipoprotein A II
0.33 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P02652
Apo AII, ApoA II, APOA2, apolipoprotein A II, Apolipoprotein A2
Rabbit
IgG
150 μL
-20°C
Primary
336
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Unconjugated
APOA2
Liquid
APOA2
Apolipoprotein A II Fusion Protein Ag9863
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human
Vidéos
FDS
Documents