missing translation for 'onlineSavingsMsg'
Learn More
Learn More
APPL2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 14294-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
Description
Required for the regulation of cell proliferation in response to extracellular signals mediated by an early endosomal compartment. Links Rab5 to nuclear signal transduction.Spécification
APPL2 | |
Polyclonal | |
Unconjugated | |
APPL2 | |
APPL2, Dip13 beta, DIP13B | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
216190, 362860, 55198 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Western Blot | |
0.27 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q8K3G9, Q8NEU8 | |
Appl2 | |
APPL2 Fusion Protein Ag5626 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |