missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ARCN1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 23843-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
Description
This gene maps in a region, which include the mixed lineage leukemia and Friend leukemia virus integration 1 genes, where multiple disease-associated chromosome translocations occur. It is an intracellular protein. Archain sequences are well conserved among eukaryotes and this protein may play a fundamental role in eukaryotic cell biology. It has similarities to heat shock proteins and clathrin-associated proteins, and may be involved in vesicle structure or trafficking.Spécification
ARCN1 | |
Polyclonal | |
Unconjugated | |
ARCN1 | |
Archain, archain 1, ARCN1, Coatomer subunit delta, COPD, Delta coat protein, Delta COP | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
213827, 372 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.22 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P48444, Q5XJY5 | |
ARCN1 | |
ARCN1 Fusion Protein Ag20836 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |