missing translation for 'onlineSavingsMsg'
Learn More

Arginase 1 Monoclonal Antibody (A1exF5), eFluor™ 450, eBioscience™, Invitrogen™

Rat Monoclonal Antibody

148.00€ - 284.00€

Spécification

Antigène Arginase 1
Clone A1exF5
Concentration 0.2 mg/mL
Applications Flow Cytometry
Classification Monoclonal
Consulter d'autres spécifications

Produits 2
Code produit Marque Quantité Prix Quantité et disponibilité  
Code produit Marque Quantité Prix Quantité et disponibilité  
15913304
Afficher les documents Détails de la promotion
Invitrogen
48369780
25 μg
148.00€
25µg
Voir les produits de remplacement
Économisez jusqu'à 
»
En stock
pour voir le stock disponible 
Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!
15923304
Afficher les documents Détails de la promotion
Thermo Scientific
48369782
100 μg
284.00€
100µg
Voir les produits de remplacement
Économisez jusqu'à 
»
En stock
pour voir le stock disponible 
Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!
Description

Description

Description: The monoclonal antibody A1exF5 recognizes both human and mouse Arginase 1, a cytosolic enzyme (Arg1). This A1exF5 clone is compatible with both, the standard intracellular protocols, and the Foxp3/Transcription Factor Staining Buffer Set. Applications Reported: This A1exF5 antibody has been reported for use in flow cytometric analysis. Applications Tested: This A1exF5 antibody has been tested by flow cytometric analysis of normal human peripheral blood cells using the Intracellular Fixation & Permeabilization Buffer Set (Product # 88-8824) and protocol. Please refer to Best Protocols: Protocol A: Two step protocol for (cytoplasmic) intracellular proteins located under the Resources Tab online. This may be used at less than or equal to 1.0 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. eFluor™ 450 is an alternative for Pacific Blue™. eFluor 450 emits at 446 nm and is excited with the violet laser line (405 nm). Please make sure that your instrument is capable of detecting this fluorochrome. Excitation: 405 nm; Emission: 445 nm; Laser: Violet Laser

Arginase 1 Monoclonal antibody specifically detects Arginase 1 in Human, Mouse samples. It is validated for Flow Cytometry.
Spécification

Spécification

Arginase 1
0.2 mg/mL
Monoclonal
Liquid
RUO
PBS with 0.09% sodium azide; pH 7.2
AI; A-I; AI type I arginase; AI256583; Arg1; Arg-1; Arginase; arginase 1; arginase 1 liver; arginase 1, liver; arginase I; arginase, liver; Arginase1; arginase-1; HGNC:663; Liver Arginase; Liver-type arginase; PGIF; similar to arginase, type I; Type 1 Arginase; type I arginase
ARG1
IgG2a κ
Affinity chromatography
Antibody
A1exF5
Flow Cytometry
eFluor 450
Rat
Human, Mouse
P05089, Q61176
11846, 383
E.coli-derived recombinant mouse Arginase 1
Primary
4° C, store in dark, DO NOT FREEZE!
ARG1
Vidéos
FDS
Documents

Documents

Product Certifications