missing translation for 'onlineSavingsMsg'
Learn More

Arginase 1 Monoclonal Antibody (A1exF5), Alexa Fluor™ 700, eBioscience™, Invitrogen™

Rat Monoclonal Antibody

Marque:  Affymetrix eBioscience 56-3697-80

90.00 EUR valable jusqu'au 2024-03-29
Utilisez le code promo "21615" pour bénéficier de cette offre.



Informations supplémentaires : Poids : 0.01000kg

 Afficher plus de versions de ce produit

Code produit. 16371034

  • 135.00€ / 25µg
Expédition estimée: 19-04-2024
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

Description: The monoclonal antibody A1exF5 recognizes both human and mouse Arginase 1, a cytosolic enzyme (Arg1). This A1exF5 clone is compatible with both, the standard intracellular protocols, and the Foxp3/Transcription Factor Staining Buffer Set. Applications Reported: This A1exF5 antibody has been reported for use in flow cytometric analysis. Applications Tested: This A1exF5 antibody has been tested by flow cytometric analysis of stimulated mouse bone marrow cells using the Intracellular Fixation & Permeabilization Buffer Set (Product # 88-8824) and protocol. Please refer to Best Protocols: Protocol A: Two step protocol for (cytoplasmic) intracellular proteins located under the Resources Tab online. This may be used at less than or equal to 0.125 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Alexa Fluor® 700 emits at 723 nm and can be excited with the red laser (633 nm). Most instruments will require a 685 LP mirror and 710/20 filter. Please make sure that your instrument is capable of detecting this fluorochrome. Excitation: 633-647 nm; Emission: 723 nm; Laser: Red Laser

Arginase 1 Monoclonal antibody specifically detects Arginase 1 in Human, Mouse samples. It is validated for Flow Cytometry.
Spécification

Spécification

Arginase 1
Monoclonal
0.2 mg/mL
PBS with 0.09% sodium azide; pH 7.2
P05089, Q61176
ARG1
E.coli-derived Recombinant mouse Arginase 1
25 μg
Primary
Human, Mouse
Antibody
IgG2a κ
Flow Cytometry
A1exF5
Alexa Fluor 700
ARG1
AI; A-I; AI type I arginase; AI256583; Arg1; Arg-1; Arginase; arginase 1; arginase 1 liver; arginase 1, liver; arginase I; arginase, liver; Arginase1; arginase-1; HGNC:663; Liver Arginase; Liver-type arginase; PGIF; similar to arginase, type I; Type 1 Arginase; type I arginase
Rat
Affinity chromatography
RUO
11846, 383
4° C, store in dark, DO NOT FREEZE!
Liquid
Suggestions de produits

Suggestions de produits

Vidéos
FDS
Documents

Documents

Certificats
Promotions

Promotions