missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ARH3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 16504-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
Description
ADPRHL2 is a member of the ADP-ribosylglycohydrolase family. The enzyme catalyzes the removal of ADP-ribose from ADP-ribosylated proteins. This enzyme localizes to the mitochondria, in addition to the nucleus and cytoplasm.Spécification
ARH3 | |
Polyclonal | |
Unconjugated | |
ADPRHL2 | |
ADP ribosylhydrolase 3, ADP ribosylhydrolase like 2, ADPRHL2, ARH3 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
100206, 362600, 54936 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.18 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q8CG72, Q9NX46 | |
Adprhl2 | |
ARH3 Fusion Protein Ag9683 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |