Learn More
ARHGAP30 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marque: Thermo Scientific PA557604
Code nomenclature Nacres: NA.41
Informations supplémentaires : Poids : 0.02000kg
Description
Immunogen sequence: ANPCPRPGRL DGTPGERAWG SRASRSSWRN GGSLSFDAAV ALARDRQRTE AQGVRRTQTC TEGGDYCLIP RTSPCSMISA HSPRPLSCLE LPSEGAEGSG SRSR Highest antigen sequence identity to the following orthologs: Mouse - 81%, Rat - 81%.
ARHGAP30 is GTPase activator for the Rho-type GTPases by converting them to an inactive GDP-bound state.Spécification
ARHGAP30 | |
Polyclonal | |
Unconjugated | |
ARHGAP30 | |
6030405P05Rik; ARHGAP30; Gm102; mFLJ00267; RGD1561419; Rho GTPase activating protein 30; rho GTPase-activating protein 30; Rho-type GTPase-activating protein 30 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
257106 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry | |
0.3 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q7Z6I6 | |
ARHGAP30 | |
Recombinant protein corresponding to Human ARHGAP30 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only