missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ARPM1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 27580-1-AP-20UL
Informations supplémentaires : Poids : 0.00010kg
Description
ACTRT3 is a protein coding gene.Spécification
ARPM1 | |
0.27 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q8BXF8, Q9BYD9 | |
actin related protein M1, ARPM1 | |
Rabbit | |
IgG | |
20 μL | |
-20°C | |
Primary | |
365763, 76652, 84517 |
Immunohistochemistry (Paraffin), Western Blot, Immunofluorescence, Immunocytochemistry | |
Unconjugated | |
ACTRT3 | |
Liquid | |
Actrt3 | |
ARPM1 Fusion Protein Ag25951 | |
Antigen Affinity Chromatography | |
RUO | |
Antibody | |
Polyclonal | |
Mouse, Rat, Human |