missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ASNS Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 14681-1-AP-20UL
Informations supplémentaires : Poids : 0.00010kg
Description
The protein encoded by this gene is involved in the synthesis of asparagine. This gene complements a mutation in the temperature-sensitive hamster mutant ts11, which blocks progression through the G1 phase of the cell cycle at nonpermissive temperature. There are three alternatively spliced transcript variants encoding the same protein described for this gene.Spécification
ASNS | |
Polyclonal | |
Unconjugated | |
ASNS | |
ASNS, asparagine synthetase, TS11 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
25612, 27053, 440 | |
-20°C | |
Liquid |
Western Blot, Immunofluorescence, Immunocytochemistry, Immunoprecipitation | |
0.27 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P08243, P49088, Q61024 | |
Asns | |
ASNS Fusion Protein Ag6393 | |
20 μL | |
Primary | |
Mouse, Human, Rat | |
Antibody | |
IgG |