Learn More
Gilson™ ASPEC™ Alumina-A SPE Cartridge, Bed Wt. 50 mg
Alumina cartridges contain a high-quality, polar sorbent, used the same way as silica, but with better pH stability (4 to 5.5), acidic character for useful separation of acid pigments, strong acids, phenols, sulfonic, carboxylic and amino acids
Marque:  Gilson™ 54350766
Informations supplémentaires : Poids : 0.01100kg
 Afficher plus de versions de ce produit
Code produit. 16646435
  • 236.00€ / 100 pièces
Date d’expédition estimée: 23-11-2021
Ajouter au panier
Description
Description
  • Pre-capped cartridges are ASPEC™ instrument-ready
  • Automation delivers consistent flow rates
    • Provide loading capacity, accurate elution properties and reproducible separation with low solvent usage
    • Cartridges available uncapped for manual extractions
Spécification
Spécification
SPE cartridge
Yes
SPE
Alumina-A
Automated extractions
50 mg
Alumina-A
ASPEC™
1 mL