Learn More
Gilson™ ASPEC™ Alumina-A SPE Cartridge, Bed Wt. 500 mg
Alumina cartridges contain a high-quality, polar sorbent, used the same way as silica, but with better pH stability (4 to 5.5), acidic character for useful separation of acid pigments, strong acids, phenols, sulfonic, carboxylic and amino acids
Marque:  Gilson™ 54350770
Informations supplémentaires : Poids : 0.01100kg
 Afficher plus de versions de ce produit
Code produit. 16626445
  • 204.00€ / 50 pièces
Date d’expédition estimée: 16-11-2021
Ajouter au panier
Description
Description
  • Pre-capped cartridges are ASPEC™ instrument-ready
  • Automation delivers consistent flow rates
    • Provide loading capacity, accurate elution properties and reproducible separation with low solvent usage
    • Cartridges available uncapped for manual extractions
Spécification
Spécification
SPE cartridge
Yes
SPE
Alumina-A
Automated extractions
500 mg
Alumina-A
ASPEC™
6 mL