Learn More
Gilson™ ASPEC™ Alumina-B SPE Cartridge, Bed Wt. 50 mg
Alumina cartridges contain a high-quality, polar sorbent, used the same way as silica, but with better pH stability (8 to 9.5), basic character makes it more selective for basic and neutral compounds
Marque:  Gilson™ 54350780
Informations supplémentaires : Poids : 0.01100kg
 Afficher plus de versions de ce produit
Code produit. 16626465
  • 236.00€ / 100 pièces
Date d’expédition estimée: 19-11-2021
Ajouter au panier
Description
Description
  • Pre-capped cartridges are ASPEC™ instrument-ready
  • Automation delivers consistent flow rates
    • Provide loading capacity, accurate elution properties and reproducible separation with low solvent usage
    • Cartridges available uncapped for manual extractions
Spécification
Spécification
SPE cartridge
Yes
SPE
Alumina-B
Automated extractions
50 mg
Alumina-B
ASPEC™
1 mL