Learn More
Gilson™ ASPEC™ Alumina-N SPE Cartridge, Bed Wt. 200 mg
Alumina cartridges contain a high-quality, polar sorbent, used the same way as silica, but with better pH stability (6 to 7.5), neutral character provides good retention of aromatic compounds, aliphatic amines and compounds with electronegative functions
Marque:  Gilson™ 54350775NC
Informations supplémentaires : Poids : 0.01100kg
 Afficher plus de versions de ce produit
Code produit. 16636455
  • 151.00€ / 50 pièces
Date d’expédition estimée: 22-11-2021
Ajouter au panier
Description
Description
  • Pre-capped cartridges are ASPEC™ instrument-ready
  • Automation delivers consistent flow rates
    • Provide loading capacity, accurate elution properties and reproducible separation with low solvent usage
    • Cartridges available uncapped for manual extractions
Spécification
Spécification
SPE cartridge
No
SPE
Alumina-N
Manual extractions
200 mg
Alumina-N
ASPEC™
3 mL