Learn More
Gilson™ ASPEC™ C18 Nec SPE 96-well Plate
C18 cartridges contain a high-quality non-polar sorbent, provide consistent flow rates, high loading capacity, accurate elution properties and reproducible separation with low solvent usage
Marque:  Gilson™ 54350885NC
Informations supplémentaires : Poids : 0.01100kg
 Afficher plus de versions de ce produit
Code produit. 16646665
  • 820.00€ / 1 pièce
Date d’expédition estimée: 19-11-2021
Ajouter au panier
Description
Description
  • ASPEC™ pre-capped cartridges are ASPEC™ instrument-ready, saving your time and money by eliminating manual capping.
  • Provide high loading capacity, accurate elution properties and reproducible separation with low solvent usage
  • Available uncapped for manual extractions
Spécification
Spécification
SPE 96-Well Plate
No
SPE
C18 NEC
50 mg
C18 NEC
ASPEC™
2 mL