Learn More
Gilson™ ASPEC™ C18 Nec SPE 96-well Plate
C18 cartridges contain a high-quality non-polar sorbent, provide consistent flow rates, high loading capacity, accurate elution properties and reproducible separation with low solvent usage
sur devis
Spécification
Volume (métrique) 2 ml
Consulter d'autres spécifications
Description
Description
  • ASPEC™ pre-capped cartridges are ASPEC™ instrument-ready, saving your time and money by eliminating manual capping.
  • Provide high loading capacity, accurate elution properties and reproducible separation with low solvent usage
  • Available uncapped for manual extractions
Spécification
Spécification
2 ml