Learn More
Gilson™ ASPEC™ C18 NEC SPE Cartridge, Bed Wt. 50 mg
C18 NEC cartridges contain a high-quality, non-polar sorbent, higher retention and polarity selectivity of basic compounds compared to C18 PLUS cartridges
Marque:  Gilson™ 54350715
Informations supplémentaires : Poids : 0.01100kg
 Afficher plus de versions de ce produit
Code produit. 16626335
  • 271.00€ / 100 pièces
Date d’expédition estimée: 23-11-2021
Ajouter au panier
Description
Description
  • Pre-capped cartridges are ASPEC™ instrument-ready
  • Automation delivers consistent flow rates
    • Provide loading capacity, accurate elution properties and reproducible separation with low solvent usage
    • Cartridges available uncapped for manual extractions
Spécification
Spécification
SPE cartridge
Yes
SPE
C18 NEC
Automated extractions
50 mg
C18
ASPEC™
1 mL