Learn More
Gilson™ ASPEC™ C18 PLUS SPE Cartridge, Bed Wt. 50 mg
C18 PLUS silica based cartridges contain a high-quality non-polar hydrophobic sorbent, for extracting organic compounds from aqueous solutions (like contaminants from water, drugs, metabolites) and physiological fluids
Marque:  Gilson™ 54350700NC
Informations supplémentaires : Poids : 0.01100kg
 Afficher plus de versions de ce produit
Code produit. 16636305
  • 212.00€ / 100 pièces
Date d’expédition estimée: 23-11-2021
Ajouter au panier
Description
Description
  • Pre-capped cartridges are ASPEC™ instrument-ready
  • Automation delivers consistent flow rates
    • Provide loading capacity, accurate elution properties and reproducible separation with low solvent usage
    • Cartridges available uncapped for manual extractions
Spécification
Spécification
SPE cartridge
No
ASPEC™
1 mL
50 mg
C18
C18 PLUS
Manual extractions