Learn More
Gilson™ ASPEC™ C18 WPD SPE 96-well Plate
Silica based ASPEC™ C18 WPD solid phase extraction (SPE) cartridges contain a high-quality non-polar, strongly hydrophobic sorbent, similar to ASPEC™ C18 PLUS solid phase with the ability to accommodate larger molecules and untreated matrices.
Marque:  Gilson™ 54350883NC
Informations supplémentaires : Poids : 0.01100kg
 Afficher plus de versions de ce produit
Code produit. 16626665
  • 820.00€ / 1 pièce
Date d’expédition estimée: 24-11-2021
Ajouter au panier
Description
Description
  • ASPEC™ pre-capped cartridges are ASPEC™ instrument-ready, saving your time and money by eliminating manual capping
  • Provide high loading capacity, accurate elution properties and reproducible separation with low solvent usage
  • Available uncapped for manual extractions
Spécification
Spécification
50 mg
ASPEC™
2 mL
No
C18 WPD