Learn More
Gilson™ ASPEC™ C18 WPD SPE Cartridge, Bed Wt. 200 mg
C18 WPD cartridges contain a high-quality non-polar strongly hydrophobic sorbent, like C18 PLUS but can accommodate larger molecules and untreated matrices
Marque:  Gilson™ 54350710NC
Informations supplémentaires : Poids : 0.01100kg
 Afficher plus de versions de ce produit
Code produit. 16636325
  • 171.00€ / 50 pièces
Date d’expédition estimée: 24-11-2021
Ajouter au panier
Description
Description
  • Pre-capped cartridges are ASPEC™ instrument-ready
  • Automation delivers consistent flow rates
    • Provide loading capacity, accurate elution properties and reproducible separation with low solvent usage
    • Cartridges available uncapped for manual extractions
Spécification
Spécification
SPE cartridge
No
SPE
C18 WPD
Manual extractions
200 mg
C18
ASPEC™
3 mL