Learn More
Gilson™ ASPEC™ C4 Nec SPE 96-well Plate
Silica based C4 cartridges contain a high-quality, non-polar, moderately hydrophobic sorbent, for overcoming to strong retention of non-polar analytes on C18 or C8 sorbent
Marque:  Gilson™ 54350922NC
Informations supplémentaires : Poids : 0.01100kg
 Afficher plus de versions de ce produit
Code produit. 16616705
  • 889.00€ / 1 pièce
Date d’expédition estimée: 18-11-2021
Ajouter au panier
Description
Description
  • ASPEC™ pre-capped cartridges are ASPEC™ instrument-ready, saving your time and money by eliminating manual capping.
  • Provide high loading capacity, accurate elution properties and reproducible separation with low solvent usage
  • Available uncapped for manual extractions
Spécification
Spécification
SPE 96-Well Plate
No
SPE
C4 NEC
100 mg
C4 NEC
ASPEC™
2 mL