Learn More
Gilson™ ASPEC™ C4 SPE Cartridge, Bed Wt. 500 mg
Silica based C4 cartridges contain a high-quality, non-polar, moderately hydrophobic sorbent, for overcoming to strong retention of non-polar analytes on C18 or C8 sorbent
Marque:  Gilson™ 54350511
Informations supplémentaires : Poids : 0.01100kg
 Afficher plus de versions de ce produit
Code produit. 16626195
  • 228.00€ / 50 pièces
Date d’expédition estimée: 16-11-2021
Ajouter au panier
Description
Description
  • Pre-capped cartridges are ASPEC™ instrument-ready
  • Automation delivers consistent flow rates
    • Provide loading capacity, accurate elution properties and reproducible separation with low solvent usage
    • Cartridges available uncapped for manual extractions
Spécification
Spécification
SPE cartridge
Yes
SPE
C4
Automated extractions
500 mg
C4
ASPEC™
6 mL