Learn More
Gilson™ ASPEC™ C8 NEC SPE Cartridge, Bed Wt. 200 mg
Silica based C8 NEC cartridges contain a high-quality, non-polar and moderate hydrophobicity sorbent, for extraction of extremely non-polar compounds (PAH, vitamins D, oil, greasy compounds)
Marque:  Gilson™ 54350725NC
Informations supplémentaires : Poids : 0.01100kg
 Afficher plus de versions de ce produit
Code produit. 16636355
  • 145.00€ / 50 pièces
Date d’expédition estimée: 18-11-2021
Ajouter au panier
Description
Description
  • Pre-capped cartridges are ASPEC™ instrument-ready
  • Automation delivers consistent flow rates
    • Provide loading capacity, accurate elution properties and reproducible separation with low solvent usage
    • Cartridges available uncapped for manual extractions
Spécification
Spécification
SPE cartridge
No
SPE
C8 NEC
Manual extractions
200 mg
C8
ASPEC™
3 mL