Learn More
Gilson™ ASPEC™ C8/SAX-2 NEC SPE Cartridge, Bed Wt. 100 mg
Silica based C8/SAX-2 NEC cartridges contain high-quality sorbent, for extracting acidic compounds from aqueous solutions (typically drugs and metabolites from physiological fluids)
Marque:  Gilson™ 54350795NC
Informations supplémentaires : Poids : 0.01100kg
 Afficher plus de versions de ce produit
Code produit. 16636495
  • 225.00€ / 100 pièces
Date d’expédition estimée: 24-11-2021
Ajouter au panier
Description
Description
  • Pre-capped cartridges are ASPEC™ instrument-ready
  • Automation delivers consistent flow rates
    • Provide loading capacity, accurate elution properties and reproducible separation with low solvent usage
    • Cartridges available uncapped for manual extractions
Spécification
Spécification
SPE cartridge
No
SPE
C8/SAX-2 NEC
Manual extractions
100 mg
C8/SAX-2
ASPEC™
1 mL