Learn More
Gilson™ ASPEC™ C8 SPE Cartridge, Bed Wt. 500 mg
C8 cartridges contain a high-quality, non-polar and moderately hydrophobicity sorbent, for extraction of non-polar compounds in aqueous solutions like pharmaceutical drugs, PAH in biological fluids (plasma, serum, urine) or herbicides in water
Marque:  Gilson™ 54350516NC
Informations supplémentaires : Poids : 0.01100kg
 Afficher plus de versions de ce produit
Code produit. 16636205
  • 159.00€ / 50 pièces
Date d’expédition estimée: 23-11-2021
Ajouter au panier
Description
Description
  • Pre-capped cartridges are ASPEC™ instrument-ready
  • Automation delivers consistent flow rates
    • Provide loading capacity, accurate elution properties and reproducible separation with low solvent usage
    • Cartridges available uncapped for manual extractions
Spécification
Spécification
SPE cartridge
No
SPE
C8
Manual extractions
500 mg
C8
ASPEC™
3 mL