Learn More
Gilson™ ASPEC™ CleanENVI SPE Cartridge, Bed Wt. 500 mg
Silica based CleanENVI cartridges contain a high-quality, certified sorbent against extractables and leachables, designed to extract typical environmental contaminants (PAHs, pesticides, herbicides from water) according to EPA regulations
Marque:  Gilson™ 54350825NC
Informations supplémentaires : Poids : 0.01100kg
 Afficher plus de versions de ce produit
Code produit. 16636555
  • 159.00€ / 50 pièces
Date d’expédition estimée: 16-11-2021
Ajouter au panier
Description
Description
  • Pre-capped cartridges are ASPEC™ instrument-ready
  • Automation delivers consistent flow rates
    • Provide loading capacity, accurate elution properties and reproducible separation with low solvent usage
    • Cartridges available uncapped for manual extractions
Spécification
Spécification
SPE cartridge
No
SPE
CleanENVI
Manual extractions
500 mg
CleanENVI
ASPEC™
3 mL