Learn More
Gilson™ ASPEC™ Cyano SPE 96-well Plate
Cyano cartridges contain a high-quality, moderately polar sorbent with polar and hydrophobic characters, used as less polar alternative in normal phase mode (compared to silica sorbent) or less hydrophobic alternative in reversed phase mode (compared to C8 and C18 PLUS sorbents)
sur devis
Spécification
Volume (métrique) 2 ml
Consulter d'autres spécifications
Description
Description
  • ASPEC™ pre-capped cartridges are ASPEC™ instrument-ready, saving your time and money by eliminating manual capping.
  • Provide high loading capacity, accurate elution properties and reproducible separation with low solvent usage
  • Available uncapped for manual extractions
Spécification
Spécification
2 ml