Learn More
Gilson™ ASPEC™ Cyano SPE Cartridge, Bed Wt. 500 mg
Cyano cartridges contain a high-quality, moderately polar sorbent with polar and hydrophobic characters, used as less polar alternative in normal phase mode (compared to silica sorbent) or less hydrophobic alternative in reversed phase mode (compared to C8 and C18 PLUS sorbents)
Marque:  Gilson™ 54350741
Informations supplémentaires : Poids : 0.01100kg
 Afficher plus de versions de ce produit
Code produit. 16646385
  • 208.00€ / 50 pièces
Date d’expédition estimée: 19-11-2021
Ajouter au panier
Description
Description
  • Pre-capped cartridges are ASPEC™ instrument-ready
  • Automation delivers consistent flow rates
    • Provide loading capacity, accurate elution properties and reproducible separation with low solvent usage
    • Cartridges available uncapped for manual extractions
Spécification
Spécification
SPE cartridge
Yes
SPE
Cyano
Automated extractions
500 mg
Cyano
ASPEC™
6 mL