Learn More
Gilson™ ASPEC™ Cyclohexyl SPE 96-well Plate
Silica based cyclohexyl cartridges contain a high-quality, non-polar sorbent, for extracting compounds from aqueous solutions with different selectivity compared to C8 and C18 PLUS sorbents
Marque:  Gilson™ 54350923NC
Informations supplémentaires : Poids : 0.01100kg
 Afficher plus de versions de ce produit
Code produit. 16626705
  • 820.00€ / 1 pièce
Date d’expédition estimée: 23-11-2021
Ajouter au panier
Description
Description
  • ASPEC™ pre-capped cartridges are ASPEC™ instrument-ready, saving your time and money by eliminating manual capping.
  • Provide high loading capacity, accurate elution properties and reproducible separation with low solvent usage
  • Available uncapped for manual extractions
Spécification
Spécification
SPE 96-Well Plate
No
SPE
Cyclohexyl
50 mg
Cyclohexyl
ASPEC™
2 mL