Learn More
Gilson™ ASPEC™ Cyclohexyl SPE Cartridge, Bed Wt. 200 mg
Silica based cyclohexyl cartridges contain a high-quality, non-polar sorbent, for extracting compounds from aqueous solutions with different selectivity compared to C8 and C18 PLUS sorbents
Marque:  Gilson™ 54350835NC
Informations supplémentaires : Poids : 0.01100kg
 Afficher plus de versions de ce produit
Code produit. 16636575
  • 166.00€ / 50 pièces
Date d’expédition estimée: 19-11-2021
Ajouter au panier
Description
Description
  • Pre-capped cartridges are ASPEC™ instrument-ready
  • Automation delivers consistent flow rates
    • Provide loading capacity, accurate elution properties and reproducible separation with low solvent usage
    • Cartridges available uncapped for manual extractions
Spécification
Spécification
SPE cartridge
No
SPE
Cyclohexyl
Manual extractions
200 mg
Cyclohexyl
ASPEC™
3 mL