Learn More
Gilson™ ASPEC™ Cyclohexyl SPE Cartridge, Bed Wt. 500 mg
Silica based cyclohexyl cartridges contain a high-quality, non-polar sorbent, for extracting compounds from aqueous solutions with different selectivity compared to C8 and C18 PLUS sorbents
Marque:  Gilson™ 54350836
Informations supplémentaires : Poids : 0.01100kg
 Afficher plus de versions de ce produit
Code produit. 16646575
  • 225.00€ / 50 pièces
Date d’expédition estimée: 22-11-2021
Ajouter au panier
Description
Description
  • Pre-capped cartridges are ASPEC™ instrument-ready
  • Automation delivers consistent flow rates
    • Provide loading capacity, accurate elution properties and reproducible separation with low solvent usage
    • Cartridges available uncapped for manual extractions
Spécification
Spécification
SPE cartridge
Yes
SPE
Cyclohexyl
Automated extractions
500 mg
Cyclohexyl
ASPEC™
3 mL