Learn More
Gilson™ ASPEC™ Diol NEC SPE Cartridge, Bed Wt. 50 mg
Silica based diol NEC cartridges contain a high-quality, moderate polar sorbent with neutral character, for extracting polar compounds and structural isomers from non-polar solvents
Marque:  Gilson™ 54350745
Informations supplémentaires : Poids : 0.01100kg
 Afficher plus de versions de ce produit
Code produit. 16626395
  • 250.00€ / 100 pièces
Date d’expédition estimée: 23-11-2021
Ajouter au panier
Description
Description
  • Pre-capped cartridges are ASPEC™ instrument-ready
  • Automation delivers consistent flow rates
    • Provide loading capacity, accurate elution properties and reproducible separation with low solvent usage
    • Cartridges available uncapped for manual extractions
Spécification
Spécification
SPE cartridge
Yes
SPE
Diol NEC
Automated extractions
50 mg
Diol
ASPEC™
1 mL