Learn More
Gilson™ ASPEC™ DVB SPE Cartridge, Bed Wt. 200 mg
Polymer based DVB cartridges contain a high-quality, hydrophobic sorbent from polystyrene-divinylbenzene copolymer, high hydrophobicity, for use as a reversed-phase for extraction of neutral, acidic and basic compounds in biological and viscous matrices
Marque:  Gilson™ 54350851
Informations supplémentaires : Poids : 0.01100kg
 Afficher plus de versions de ce produit
Code produit. 16646605
  • 263.00€ / 30 pièces
Date d’expédition estimée: 23-11-2021
Ajouter au panier
Description
Description
  • Pre-capped cartridges are ASPEC™ instrument-ready
  • Automation delivers consistent flow rates
    • Provide loading capacity, accurate elution properties and reproducible separation with low solvent usage
    • Cartridges available uncapped for manual extractions
Spécification
Spécification
SPE cartridge
Yes
SPE
DVB
Automated extractions
200 mg
DVB
ASPEC™
6 mL