Learn More
Gilson™ ASPEC™ Florisil PR SPE 96-well Plate
Magnesium based florisil PR cartridges contain a high-quality, polar sorbent with pesticide residue analysis grade, for isolating polar to moderate polar compounds for non-polar matrices (recommended for isolation of chlorinated pesticides, polychlorinated biphenyl and polysaccharides)
sur devis
Spécification
Volume (métrique) 2 ml
Consulter d'autres spécifications
Description
Description
  • ASPEC™ pre-capped cartridges are ASPEC™ instrument-ready, saving your time and money by eliminating manual capping.
  • Provide high loading capacity, accurate elution properties and reproducible separation with low solvent usage
  • Available uncapped for manual extractions
Spécification
Spécification
2 ml