Learn More
Gilson™ ASPEC™ Florisil PR SPE Cartridge, Bed Wt. 100 mg
Magnesium based florisil PR cartridges contain a high-quality, polar sorbent with pesticide residue analysis grade, for isolating polar to moderate polar compounds for non-polar matrices (recommended for isolation of chlorinated pesticides, polychlorinated biphenyl and polysaccharides)
Marque:  Gilson™ 54350760NC
Informations supplémentaires : Poids : 0.01100kg
 Afficher plus de versions de ce produit
Code produit. 16636425
  • 206.00€ / 100 pièces
Date d’expédition estimée: 22-11-2021
Ajouter au panier
Description
Description
  • Pre-capped cartridges are ASPEC™ instrument-ready
  • Automation delivers consistent flow rates
    • Provide loading capacity, accurate elution properties and reproducible separation with low solvent usage
    • Cartridges available uncapped for manual extractions
Spécification
Spécification
SPE cartridge
No
SPE
Florisil PR
Manual extractions
100 mg
Florisil
ASPEC™
1 mL