Learn More
Gilson™ ASPEC™ Florisil SPE 96-well Plate
Magnesium based florisil cartridges contain a high-quality, polar sorbent, for isolating polar to moderate polar compounds from non-polar matrices
Marque:  Gilson™ 54350902NC
Informations supplémentaires : Poids : 0.01100kg
 Afficher plus de versions de ce produit
Code produit. 16616685
  • 820.00€ / 1 pièce
Date d’expédition estimée: 24-11-2021
Ajouter au panier
Description
Description
  • ASPEC™ pre-capped cartridges are ASPEC™ instrument-ready, saving your time and money by eliminating manual capping.
  • Provide high loading capacity, accurate elution properties and reproducible separation with low solvent usage
  • Available uncapped for manual extractions
Spécification
Spécification
SPE 96-Well Plate
No
SPE
Florisil
100 mg
Florisil
ASPEC™
2 mL