Learn More
Gilson™ ASPEC™ Florisil SPE Cartridge, Bed Wt. 500 mg
Magnesium based florisil cartridges contain a high-quality, polar sorbent, for isolating polar to moderate polar compounds from non-polar matrices
Marque:  Gilson™ 54350755NC
Informations supplémentaires : Poids : 0.01100kg
 Afficher plus de versions de ce produit
Code produit. 16636415
  • 168.00€ / 50 pièces
Date d’expédition estimée: 18-11-2021
Ajouter au panier
Description
Description
  • Pre-capped cartridges are ASPEC™ instrument-ready
  • Automation delivers consistent flow rates
    • Provide loading capacity, accurate elution properties and reproducible separation with low solvent usage
    • Cartridges available uncapped for manual extractions
Spécification
Spécification
SPE cartridge
No
SPE
Florisil
Manual extractions
500 mg
Florisil
ASPEC™
3 mL