Learn More
Gilson™ ASPEC™ HLB SPE Cartridge, Bed Wt. 30 mg
Polymer based HLB cartridges contain a high-quality, wettable copolymer with hydrophilic lipophilic balance, demonstrating strong retention for neutral, acidic and basic compounds
Marque:  Gilson™ 54350561NC
Informations supplémentaires : Poids : 0.01100kg
 Afficher plus de versions de ce produit
Code produit. 16636295
  • 281.00€ / 100 pièces
Date d’expédition estimée: 16-11-2021
Ajouter au panier
Description
Description
  • Pre-capped cartridges are ASPEC™ instrument-ready
  • Automation delivers consistent flow rates
    • Provide loading capacity, accurate elution properties and reproducible separation with low solvent usage
    • Cartridges available uncapped for manual extractions
Spécification
Spécification
SPE cartridge
No
SPE
HLB
Manual extractions
30 mg
HLB
ASPEC™
1 mL