Learn More
Gilson™ ASPEC™ PCB SPE Cartridge, Bed Wt. 500 mg
Dual layer PCB cartridges contain a high-quality sorbent, combines strong cation exchange with high polar functionalities targeted toward extraction of PCB from waste oil after hexane extraction
Marque:  Gilson™ 54350811
Informations supplémentaires : Poids : 0.01100kg
 Afficher plus de versions de ce produit
Code produit. 16646525
  • 216.00€ / 50 pièces
Date d’expédition estimée: 19-11-2021
Ajouter au panier
Description
Description
  • Pre-capped cartridges are ASPEC™ instrument-ready
  • Automation delivers consistent flow rates
    • Provide loading capacity, accurate elution properties and reproducible separation with low solvent usage
    • Cartridges available uncapped for manual extractions
Spécification
Spécification
SPE cartridge
Yes
SPE
PCB
Automated extractions
500 mg
PCB
ASPEC™
3 mL