Learn More
Gilson™ ASPEC™ Phenyl SPE Cartridge, Bed Wt. 50 mg
Silica based phenyl cartridges contain a high-quality, moderately hydrophobic and non-polar sorbent, for extracting aromatic and complex natural compounds from aqueous solutions
Marque:  Gilson™ 54350730NC
Informations supplémentaires : Poids : 0.01100kg
 Afficher plus de versions de ce produit
Code produit. 16636365
  • 225.00€ / 100 pièces
Date d’expédition estimée: 22-11-2021
Ajouter au panier
Description
Description
  • Pre-capped cartridges are ASPEC™ instrument-ready
  • Automation delivers consistent flow rates
    • Provide loading capacity, accurate elution properties and reproducible separation with low solvent usage
    • Cartridges available uncapped for manual extractions
Spécification
Spécification
SPE cartridge
No
SPE
Phenyl
Manual extractions
50 mg
Phenyl
ASPEC™
1 mL