Learn More
Gilson™ ASPEC™ SAX Nec SPE 96-well Plate
Silica based SAX NEC cartridges contain a high-quality, strong anion exchanger from trimethylammonium chloride bonded sorbent, for extracting weak acid compounds (pKa 3 to 5) from aqueous and organic solutions
Marque:  Gilson™ 54350905NC
Informations supplémentaires : Poids : 0.01100kg
 Afficher plus de versions de ce produit
Code produit. 16646685
  • 820.00€ / 1 pièce
Date d’expédition estimée: 23-11-2021
Ajouter au panier
Description
Description
  • ASPEC™ pre-capped cartridges are ASPEC™ instrument-ready, saving your time and money by eliminating manual capping.
  • Provide high loading capacity, accurate elution properties and reproducible separation with low solvent usage
  • Available uncapped for manual extractions
Spécification
Spécification
SPE 96-Well Plate
No
SPE
SAX NEC
50 mg
SAX NEC
ASPEC™
2 mL