Learn More
Gilson™ ASPEC™ SAX NEC SPE Cartridge, Bed Wt. 200 mg
Silica based SAX NEC cartridges contain a high-quality, strong anion exchanger from trimethylammonium chloride bonded sorbent, for extracting weak acid compounds (pKa 3 to 5) from aqueous and organic solutions
Marque:  Gilson™ 54350527
Informations supplémentaires : Poids : 0.01100kg
 Afficher plus de versions de ce produit
Code produit. 16646225
  • 167.00€ / 50 pièces
Date d’expédition estimée: 19-11-2021
Ajouter au panier
Description
Description
  • Pre-capped cartridges are ASPEC™ instrument-ready
  • Automation delivers consistent flow rates
    • Provide loading capacity, accurate elution properties and reproducible separation with low solvent usage
    • Cartridges available uncapped for manual extractions
Spécification
Spécification
SPE cartridge
Yes
SPE
SAX NEC
Automated extractions
200 mg
SAX
ASPEC™
3 mL