Learn More
Gilson™ ASPEC™ SCX-2/SAX NEC SPE Cartridge, Bed Wt. 500 mg
Silica based mixed-mode SCX-2/SAX NEC cartridges contain high-quality propylsulfonic acid/trimethylammonium chloride bonded sorbents, for extracting acidic and basic molecules from non-ionizable molecules (cathecolamines from biofluids)
Marque:  Gilson™ 54350805NC
Informations supplémentaires : Poids : 0.01100kg
 Afficher plus de versions de ce produit
Code produit. 16636515
  • 197.00€ / 50 pièces
Date d’expédition estimée: 23-11-2021
Ajouter au panier
Description
Description
  • Pre-capped cartridges are ASPEC™ instrument-ready
  • Automation delivers consistent flow rates
    • Provide loading capacity, accurate elution properties and reproducible separation with low solvent usage
    • Cartridges available uncapped for manual extractions
Spécification
Spécification
SPE cartridge
No
SPE
SCX-2/SAX NEC
Manual extractions
500 mg
SCX-2/SAX
ASPEC™
6 mL