Learn More
Gilson™ ASPEC™ SCX-2 SPE Cartridge, Bed Wt. 500 mg
Silica based SCX-2 cartridges contain high-quality, strong cation exchange from a propylsulfonic acid bonded sorbents, slightly less acidic than SCX, maintain negative charge throughout pH scale, for extracting weak base compounds
Marque:  Gilson™ 54350546
Informations supplémentaires : Poids : 0.01100kg
 Afficher plus de versions de ce produit
Code produit. 16626265
  • 204.00€ / 50 pièces
Date d’expédition estimée: 22-11-2021
Ajouter au panier
Description
Description
  • Pre-capped cartridges are ASPEC™ instrument-ready
  • Automation delivers consistent flow rates
    • Provide loading capacity, accurate elution properties and reproducible separation with low solvent usage
    • Cartridges available uncapped for manual extractions
Spécification
Spécification
SPE cartridge
Yes
SPE
SCX-2
Automated extractions
500 mg
SCX-2
ASPEC™
3 mL