Learn More
Gilson™ ASPEC™ SCX SPE 96-well Plate
Silica based SCX cartridges contain high-quality, strong cation exchanger from a Tosic acid bonded sorbent, maintains a negative change throughout pH scale, for isolating weak basic compounds (7 < pKa < 10) in aqueous samples
sur devis
Description
- ASPEC™ pre-capped cartridges are ASPEC™ instrument-ready, saving your time and money by eliminating manual capping.
- Provide high loading capacity, accurate elution properties and reproducible separation with low solvent usage
- Available uncapped for manual extractions
Specifications
2 ml |