Learn More
Gilson™ ASPEC™ SCX SPE Cartridge, Bed Wt. 50 mg
Silica based SCX cartridges contain high-quality, strong cation exchanger from a Tosic acid bonded sorbent, maintains a negative change throughout pH scale, for isolating weak basic compounds (7 < pKa < 10) in aqueous samples
Marque:  Gilson™ 54350537
Informations supplémentaires : Poids : 0.01100kg
 Afficher plus de versions de ce produit
Code produit. 16646245
  • 292.00€ / 100 pièces
Date d’expédition estimée: 23-11-2021
Ajouter au panier
Description
Description
  • Pre-capped cartridges are ASPEC™ instrument-ready
  • Automation delivers consistent flow rates
    • Provide loading capacity, accurate elution properties and reproducible separation with low solvent usage
    • Cartridges available uncapped for manual extractions
Spécification
Spécification
SPE cartridge
Yes
SPE
SCX
Automated extractions
50 mg
SCX
ASPEC™
1 mL