Learn More
Gilson™ ASPEC™ Silica SPE Cartridge, Bed Wt. 50 mg
Silica cartridges contain a high-quality, high polar sorbent with a slightly acidic character, for extracting polar compounds from non-polar solvents through hydrogen bonding
Marque:  Gilson™ 54350501
Informations supplémentaires : Poids : 0.01100kg
 Afficher plus de versions de ce produit
Code produit. 16626175
  • 217.00€ / 100 pièces
Date d’expédition estimée: 19-11-2021
Ajouter au panier
Description
Description
  • Pre-capped cartridges are ASPEC™ instrument-ready
  • Automation delivers consistent flow rates
    • Provide loading capacity, accurate elution properties and reproducible separation with low solvent usage
    • Cartridges available uncapped for manual extractions
Spécification
Spécification
SPE cartridge
Yes
SPE
Silica
Automated extractions
50 mg
Silica
ASPEC™
1 mL