Learn More
Gilson™ ASPEC™ WAX-2 SPE 96-well Plate
Silica based WAX-2 cartridges contain a high-quality, weak anion exchange (triethylamine) sorbent, for extracting strong acidic compounds (pKa < 3) from aqueous or organic solutions with moderate non-polar character due to alkyl chain on amino function
Marque:  Gilson™ 54350925NC
Informations supplémentaires : Poids : 0.01100kg
 Afficher plus de versions de ce produit
Code produit. 16646705
  • 820.00€ / 1 pièce
Date d’expédition estimée: 23-11-2021
Ajouter au panier
Description
Description
  • ASPEC™ pre-capped cartridges are ASPEC™ instrument-ready, saving your time and money by eliminating manual capping.
  • Provide high loading capacity, accurate elution properties and reproducible separation with low solvent usage
  • Available uncapped for manual extractions
Spécification
Spécification
SPE 96-Well Plate
No
SPE
WAX-2
50 mg
WAX-2
ASPEC™
2 mL