Learn More
Gilson™ ASPEC™ WAX SPE Cartridge, Bed Wt. 200 mg
Silica based WAX cartridges contain a high-quality, weak anion exchange (amine) sorbent, for extracting structural isomers, saccharides and strong acidic compounds (pKa < 3) from aqueous or organic solutions
Marque:  Gilson™ 54350551NC
Informations supplémentaires : Poids : 0.01100kg
 Afficher plus de versions de ce produit
Code produit. 16636275
  • 151.00€ / 50 pièces
Date d’expédition estimée: 16-11-2021
Ajouter au panier
Description
Description
  • Pre-capped cartridges are ASPEC™ instrument-ready
  • Automation delivers consistent flow rates
    • Provide loading capacity, accurate elution properties and reproducible separation with low solvent usage
    • Cartridges available uncapped for manual extractions
Spécification
Spécification
SPE cartridge
No
SPE
WAX
Manual extractions
200 mg
WAX
ASPEC™
3 mL